Spectrum Chemical Magnesium Chloride, Hexahydrate

Quality and delivery you can count on every time!
  • Your source of analytical, research, development, and production-quantity fine chemicals for nearly 40 years
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$77.60 - $202.25USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Weight (g) Description
Availability Pricing
Spectrum Chemical Magnesium Chloride, Hexahydrate, Crystal, Reagent ACS Grade; 125 g EW-80122-71 Mfr # M1035-125GM
COMPARE
ACS Reagent 125 Magnesium Chloride, Hexahydrate, Crystal, Reagent ACS Grade; 125 g
$120.75USD / Each
Spectrum Chemical Magnesium Chloride, Hexahydrate, Crystal, Reagent ACS Grade; 25 g EW-80122-76 Mfr # M1035-25GM
COMPARE
ACS Reagent 25 Magnesium Chloride, Hexahydrate, Crystal, Reagent ACS Grade; 25 g
$77.60USD / Each
Spectrum Chemical Magnesium Chloride, Hexahydrate, Crystal, Reagent ACS Grade; 500 g EW-80122-77 Mfr # M1035-500GM
COMPARE
ACS Reagent 500 Magnesium Chloride, Hexahydrate, Crystal, Reagent ACS Grade; 500 g
$136.25USD / Each
Spectrum Chemical Magnesium Chloride, Hexahydrate, Crystal, USP, EP, BP; 500 g EW-80138-72 Mfr # MA110-500GMPL
COMPARE
BP, EP, and USP 500 Magnesium Chloride, Hexahydrate, Crystal, USP, EP, BP; 500 g
$202.25USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat