Spectrum Chemical® Water, HPLC Grade
Quality and consistency your lab can count on
  • Water meets HPLC grade standards
Water, HPLC Grade, is used in most HPLC mobile phases and in sample preparation. Because it dissolves most substances and is nontoxic, it is known as a Universal Solvent. To be used for HPLC analysis, it must conform to the highest purity standards. All HPLC solvents offered by Spectrum are glass distilled, sub-micron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$137.00 - $532.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Spectrum Chemical Water, HPLC Grade, 4 L EW-80109-69 Mfr # HP902-4LT
COMPARE
7732-18-5 4000 Water, HPLC Grade, 4 L
$137.00USD / Each
Spectrum Chemical Water, HPLC Grade, 4 x 4 L EW-80109-70 Mfr # HP902-4LT-CS4
COMPARE
7732-18-5 16000 Water, HPLC Grade, 4 x 4 L
$532.50USD / Case of 4