Spex CertiPrep® Assurance® Grade Interference Check Standard 5
Multi-element standards for AA & ICP instrumentation
  • Set or confirm background correction intervals
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition)
Use these Interference Check Standards to set or confirm that the correct background correction intervals have been set for sequential ICP spectrometers and that the proper inter-element correction factors are set for simultaneous ICP spectrometers.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
$304.00 - $521.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
Multiple 125 Assurance® Grade Interference Check Standard 5 for AA & ICP in 5% HNO3; 125 mL
$304.00USD / Each
COMPARE
Multiple 500 Assurance® Grade Interference Check Standard 5 for AA & ICP in 5% HNO3; 500 mL
$521.00USD / Each