Spex CertiPrep® Chloride Single-Element Ion Anion Standard
Single element standard for ion chromatography
  • Ideal for a variety of different kinds of charged analytes
  • Manufactured under ISO ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary.
These solutions offer the highest quality IC standards available for the analytical laboratory. Spex CertiPrep®Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
$67.00 - $152.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
COMPARE
Chloride 125 Chloride Single-Element Ion Anion Standard, 100 µg/mL in H2O; 125 mL
$67.00USD / Each
COMPARE
Chloride 125 Chloride Single-Element Ion Anion Standard, 1,000 µg/mL in H2O; 125 mL
$77.00USD / Each
COMPARE
Chloride 500 Chloride Single-Element Ion Anion Standard, 100 µg/mL in H2O; 500 mL
$142.00USD / Each
COMPARE
Chloride 500 Chloride Single-Element Ion Anion Standard, 1,000 µg/mL in H2O; 500 mL
$152.00USD / Each