Spex CertiPrep® Claritas PPT® Grade ICP-MS Initial Calibration Verification Standards
Multi-element standards for ICP and ICP-MS instrumentation
  • Tune and calibrate ICP-MS instrumentation prior to analysis
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
Spex CertiPrep® Claritas PPT® grade standards are a class of inorganic Certified Reference standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.
Our Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS. Spex CertiPrep®Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.
Dilute to the concentration appropriate for the instrument with equal parts of Claritas PPT® Nitric Acid Blank and Water Blank. For preparation every two weeks, or as needed.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
$133.00 - $507.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Matrix Volume (mL) Description
Availability Pricing
COMPARE
5% HNO3/tr. Tartaric Acid 125 ICP-MS Initial Calibration Verification Standard 1 in 5% HNO3/tr. Tartaric Acid, 125 mL
$507.00USD / Each
COMPARE
2% HNO3/tr. HF 125 ICP-MS Initial Calibration Verification Standard 2, 10 µg/mL (10 ppm) in 2% HNO3/tr. HF, 125 mL
$133.00USD / Each
COMPARE
5% HNO3/tr. Tartaric Acid 125 ICP-MS Initial Calibration Verification Standard 3 in 5% HNO3/tr. Tartaric Acid, 125 mL
$455.00USD / Each