Spex CertiPrep® Ionic Strength Adjustment Buffers in H2O
Increases the accuracy of the reading with ionic strength adjustment buffers
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
Ionic strength adjuster helps reduce measurement error by ensuring that samples and standards have a similar ionic strength.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$161.00 - $191.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
COMPARE
500 5 M Sodium Nitrate (NaNo3) Ionic Strength Adjustment Buffer in H2O; 500mL
$164.00USD / Each
COMPARE
500 10 M Sodium Hydroxide (NaOH) Ionic Strength Adjustment Buffer in H2O; 500 mL
$191.00USD / Each
COMPARE
500 Low Level TISAB II Ionic Strength Adjustment Buffer in H2O; 500 mL
$161.00USD / Each