Spex CertiPrep® pH 4 Calibration Standards, NIST Traceable
Meet or exceed performance criteria with QC samples
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Method 150.1
The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness.
QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.
$18.00 - $43.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
4 S.U. 475 pH 4 QC Sample, NIST Traceable, 1 Pint; 1/EA
$18.00USD / Each
COMPARE
4 S.U. 1000 pH 4 QC Sample, NIST Traceable, 1 L; 1/EA
$43.00USD / Each