Spex CertiPrep® Propylene Oxide Organic Standards
Single-component organic standards ensure consistent results
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Single-component standards offer high-quality, consistent results while saving preparation time. Select propylene oxide standard supplied in methanol-P&T or heptane. Additional solvents include acetone, methylene chloride, hexane, benzene, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at CRMsales@spex.com or call us at 732-549-7144.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$41.00 - $150.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Element/Ion Matrix
Availability Pricing
COMPARE
1,000 µg/mL (1,000 ppm) Propylene oxide Methanol-P&T
$41.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) Propylene oxide Heptane
$150.00USD / Each