Spex CertiPrep® Ultra Pure Grade Additives for XRF, ICP & AA
Additives for XRF, ICP and AA
  • Prevent borate glass from sticking to the platinum ware
  • Ultra pure grade—99.998% purity
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
Nonwetting additives act as a releasing agent to reduce the risk of sample disk sticking to the platimum ware. This makes it easier to remove the disk after fusion, protecting your sample and equipment.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at CRMsales@spex.com or call us at 732-549-7144.
$44.00 - $322.50USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Weight (g) Description
Availability Pricing
COMPARE
125 Lithium Bromide Crystals Ultra Pure Additive; 125 g
$183.00USD / Each
COMPARE
15 Lithium Bromide Solution Ultra Pure Additive; 15 mL
$44.00USD / Each
COMPARE
15 Lithium Bromide Solution (10 pack) Ultra Pure Additive; 10 x 15 mL
$297.00USD / Each
COMPARE
1000 Lithium Carbonate Ultra Pure Additive; 1 kg
$275.00USD / Each
COMPARE
125 Lithium Fluoride Crystals Ultra Pure Additive; 125 g
$219.50USD / Each
COMPARE
15 Lithium Iodide Solution Ultra Pure Additive; 15 mL
$82.00USD / Each
COMPARE
500 Lithium Nitrate Crystals Ultra Pure Additive; 500 g
$322.50USD / Each
Showing 1- 7 of 7