Spex CertiPrep® Volatile Organics Combination Mixes
Analytical standards for drinking water, wastewater, and solid waste
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 524 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to Methanol 524.3 Revision 4 are supplied in a 24-component mix.
US EPA Method 8021 is a general analytical method for the monitoring of VOCs by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
$91.00 - $171.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Concentration Description
Availability Pricing
COMPARE
200 µg/mL (200 ppm) Volatile Organics Combination Mix (54 Components), 200 µg/mL in Methanol-P&T; 1 mL
$91.00USD / Each
COMPARE
200 µg/mL (200 ppm) Volatile Organics Combination Mix (60 Components), 200 µg/mL in Methanol-P&T; 1 mL
$166.00USD / Each
COMPARE
2,000 µg/mL (2,000 ppm) Volatile Organics Combination Mix for US EPA Methods 502.2, 524, 8021, and 8260 (54 Components), 2,000 µg/mL in Methanol-P&T; 1 mL
$126.00USD / Each
COMPARE
2,000 µg/mL (2,000 ppm) Volatile Organics Combination Mix for US EPA Methods 502.2 and 524 (54 Components), 2,000 µg/mL in Methanol-P&T; 1 mL
$171.00USD / Each