Cole-Parmer® Essentials Protects Frequent-Use Hand Soap

Reduce cross-contamination with this laboratory-grade soap
  • Effectively yet gently washes away the most common germs for better hand health
  • Contains natural moisturizers like aloe and lanolin that nourish your skin
  • pH balanced (5.3-5.4) to skin's natural acid mantle barrier
The soothing coconut-based formula with moisturizing lanolin and aloe is gentle to skin, so it cleans without drying or chapping. Used by scientists who wash their hands up to 40 times a day. Available in a 17 oz (500 mL) convenient pump dispenser or 3 oz (90 mL) pocket or purse size.
$14.00 - $14.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Format Volume (Liters) Description
Availability Pricing
COMPARE
Pump 0.5 Hand Soap, Unscented, 17 oz (500 mL) Pump
$14.50USD / Each
COMPARE
Pump 0.5 Hand Soap, Citrus Scent, 17 oz (500 mL) Pump
$14.00USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat