This product is now discontinued, View Related Products
Spex® SamplePrep Genoseal™ Microplate Sealer Accessories
Optimize performance of the Spex® SamplePrep Genoseal™ heat sealer
  • Choose from a wide range of accessories to enhance operation of the Genoseal™
These replacement and optional accessories are suitable for use with the Spex® SamplePrep Genoseal™ semi-automatic heat sealer, item 41071-39. Genoseal is sold separately.
Plate weights keep the seal flat before sealing and help prevent the plate from sticking to the heater block. The plate locator allows for the plate skirt to sit around the corresponding locator and support the bottom of the plate. Various plate adapters are available for a variety of different types of microplates and support each cell individually to ensure the seal is even.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Plate Type Description
Availability Pricing
COMPARE
Plate Adapter for 384-well PCR Plate Genoseal™ Plate Adapter for 384-well PCR Plate, Flat Top
Call For Price
COMPARE
Plate Weight for Recessed Plates Genoseal™ Plate Weight for Recessed Plates
Call For Price
COMPARE
Plate Weight for Standard and Rimless Plates Genoseal™ Plate Weight for Standard and Rimless Plates, Brass
Call For Price
COMPARE
Plate Adapter for 96-well Skirtless PCR Plate Genoseal™ Skirtless Plate Adapter, 96-well
Call For Price
COMPARE
Suitable for all microplates Heat Sealing Film, EVA, 105 µL; 100/Pk
Call For Price
COMPARE
Plate Locator for Skirted Plates Genoseal™ Plate Locator for Skirted Plates