Spex® SamplePrep Grinding Vial Sets for Freezer/Mill® Cryogenic Grinders
Everything you need to make one complete grinding vial
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
$664.00USD / Pkg of 1
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Sample Size (mL) Min Sample Size (mL)
Availability Pricing
COMPARE
PC cylinder and end plugs, PC encapsulated steel impactor 4 0.5
$343.25USD / Pkg of 1
COMPARE
PC cylinder, low-chrome steel end plugs and impactor 4 0.5
$391.25USD / Pkg of 1
COMPARE
SS cylinder, end plugs with O-ring, and impactor 4 0.5
$462.75USD / Pkg of 1
COMPARE
PC cylinders, SS end plugs and impactor 0.5 0.1
$797.00USD / Pkg of 1
COMPARE
PC cylinder, SS end plugs and impactor 4 0.5
$312.00USD / Pkg of 1
COMPARE
PC cylinder, SS end plugs and impactor 25 1
$576.50USD / Pkg of 1
COMPARE
PC cylinder, SS end plugs and impactor 50 1
$774.00USD / Pkg of 1
COMPARE
SS cylinder, end plugs with O-ring, and impactor 50 1
$780.50USD / Pkg of 1
COMPARE
PC cylinder, low-chrome steel end plugs and impactor 50 1
$854.00USD / Pkg of 1
COMPARE
PC cylinder, low-chrome steel end plugs and impactor 25 1
$664.00USD / Pkg of 1
COMPARE
PC cylinder and end plugs, PC encapsulated steel impactor 25 1
$629.00USD / Pkg of 1
Showing 1- 11 of 11