Spex® SamplePrep Katanax® K1 Prime Single-Position Electric Fusion Fluxers
Prepare fused beads for X-ray fluorescence (XRF) analysis and solutions for AA, ICP, and wet chemistry analysis
  • Fuse samples without flames
  • Throughput of 5 to 7 samples per hour
Accurately assess the elemental composition of samples with fusion fluxers. Automated control of fusion parameters including heating time and temperature, mixing time and rate, cooling time for glass disks or stirring time for solutions offer exceptional results every time. Independent heaters for crucible and mold provide optimal temperature control and homogenization. Built with user safety in mind, fluxers do not use propane or LPG so there are no pipes and no risk of gas leakage or open flames. Plus, a built-in safety shield offers added protection during the fusion process. A color LCD touch-screen interface with icons and menus makes it easy to operate. Select from five languages and use preloaded or customized fusion methods. Constant display of temperature makes it easy to monitor the progress. USB connectivity offers increased convenience and accessibility.
$21,100.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
1150 115 Single-Position Electric Fusion Fluxer, 30 mm; 115 VAC
$21,100.00USD / Each
COMPARE
1150 115 Single-Position Electric Fusion Fluxer, 32 mm; 115 VAC
$21,100.00USD / Each
COMPARE
1150 115 Single-Position Electric Fusion Fluxer, 35 mm; 115 VAC
$21,100.00USD / Each
COMPARE
1150 115 Single-Position Electric Fusion Fluxer, 40 mm; 115 VAC
$21,100.00USD / Each
COMPARE
1150 115 Single-Position Electric Fusion Fluxer, Solution; 115 VAC
$21,100.00USD / Each
COMPARE
1150 230 Single-Position Electric Fusion Fluxer, 30 mm; 230 VAC
$21,100.00USD / Each
COMPARE
1150 230 Single-Position Electric Fusion Fluxer, 32 mm; 230 VAC
$21,100.00USD / Each
COMPARE
1150 230 Single-Position Electric Fusion Fluxer, 35 mm; 230 VAC
$21,100.00USD / Each
COMPARE
1150 230 Single-Position Electric Fusion Fluxer, 40 mm; 230 VAC
$21,100.00USD / Each
COMPARE
1150 230 Single-Position Electric Fusion Fluxer, Solution; 230 VAC
$21,100.00USD / Each
Showing 1- 10 of 10