Cole-Parmer® Spex® Plastic Vials for High-Energy Mixer/Mill® Grinders
Use with high-energy Mixer/Mill® grinders to pulverize and mix samples
  • Suitable for use with Cole-Parmer® Mixer/Mill® High-Energy Ball Mills
Order methacrylate balls to use with disposable plastic vials.
$52.50 - $312.00USD / Pkg of 100
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Tube Material Volume (mL)
Availability Pricing
COMPARE
Polystyrene Grinding Vials with Slip-On Polyethylene Cap, 2.5 mL; 100/PK Polystyrene 2.5
$52.50USD / Pkg of 100
COMPARE
Polystyrene Grinding Vials with Slip-On Polyethylene Cap, 5 mL; 100/PK Polystyrene 5
$126.00USD / Pkg of 100
COMPARE
Polypropylene Grinding Vials with Attached Flip Cap, 12 mL; 100/PK Polypropylene 12
$83.00USD / Pkg of 100
COMPARE
Polypropylene Grinding Vials with Attached Flip Cap, 35 mL; 100/PK Polypropylene 35
$131.25USD / Pkg of 100
COMPARE
Polypropylene Grinding Vials with Attached Flip Cap, 75 mL; 100/PK Polypropylene 75
$143.00USD / Pkg of 100
COMPARE
Polystyrene Grinding Vials with Screw-On Cap, 135 mL; 100/PK Polystyrene 135
$312.00USD / Pkg of 100
COMPARE
Polycarbonate Grinding Vials with Slip-On Polyethylene Cap, 5 mL; 100/PK Polycarbonate 5
$99.80USD / Pkg of 100
COMPARE
Polycarbonate Grinding Vials Reinforced with Slip-On Cap, 12 mL; 100/PK Polycarbonate 12
$136.50USD / Pkg of 100
Showing 1- 8 of 8