Cole-Parmer® Spex® XP-400 X-Press® Hydraulic Laboratory Pellet Press
Press samples quickly and repetitively within minutes
  • Ideal for accurate and repetitive pressing of sample pellets for XRF and other analytical techniques
  • Automated, programmable laboratory press allows for samples to be pressed within minutes
  • Ensure safe operation with safety interlock and automatic pump shut-off safety valve
  • Compact design efficiently uses your laboratory bench space
  • Programmable, user-friendly LCD touch-screen interface
The Cole-Parmer® hydraulic laboratory pellet press is an automatic and fully programmable pellet press designed for repetitive pressing of sample pellets for XRF, IR, and other analytical techniques. This press can also be operated manually as a simple motorized press whose functions are under direct user control.
The programmable LCD touch-screen interface allows the use to program the final desired pressure, time to hold at final pressure, and interval to return back to zero pressure. Safety features include a safety interlock that holds the door of the chamber in a locked position during operation and an automatic pump shut-off safety valve to ensure the safety of users.
This 35-ton (31.8 metric ton) hydraulic laboratory pellet press accepts 13-, 31-, 35-, and 40-mm pellet die sets.
$18,126.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
115 X-Press® Hydraulic Laboratory Pellet Press, 35 Ton; 115V 60HZ
$18,126.00USD / Each
COMPARE
230 X-Press® Hydraulic Laboratory Pellet Press, 35 Ton; 230V 50HZ
$18,126.00USD / Each