Cole-Parmer® Spex® XRF Prep-Aid® Grinding Aids and Sample Binders
Prepare samples for reproducible and contamination-free results
  • Select binders from powder, tablet, or liquid form depending on your application
High-quality sample preparation is the key to achieving consistent and reliable analytical results. Powders are ideal for samples that crumble or ablate after pressing. They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks. Tablets are an efficient way to add binder to XRF sample disks. Liquid easily mixes with the sample.
$52.50 - $493.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
COMPARE
Liquid Vertrel™ XF Liquid Fluorocarbon Grinding Aid, 1 Quart
$320.00USD / Each
COMPARE
Powder Cellulose Binder Powder, <20 µm, 150 g
$63.00USD / Each
COMPARE
Powder Cellulose Binder Powder, <20 µm, 450 g
$124.00USD / Each
COMPARE
Powder UltraBind® Binder Powder, <20 µm, 150 g
$124.00USD / Each
COMPARE
Powder UltraBind® Binder Powder, <20 µm, 450 g
$312.00USD / Each
COMPARE
Tablet UltraBind® Binder Tablets, 0.5 g Each, 1500 Tablets
$493.00USD / Each
COMPARE
Tablet UltraBind® Binder Tablets, 0.5 g Each, 500 Tablets
$191.50USD / Each
COMPARE
Powder Paraffin Binder Powder, <30 µm, 150 g
$52.50USD / Each
COMPARE
Powder Paraffin Binder Powder, <30 µm, 450 g
$101.00USD / Each
Showing 1- 9 of 9