Cole-Parmer® Spex® XRF Spec-Caps® Cups
Aid in the production of pressed powder sample disks for XRF, OES, and other analytical techniques
  • Reinforces sample disks to resist chipping and breaking
  • Makes sample disks easier to remove from the pellet die
  • Minimizes contamination during sample handling
  • Protects the bore of the pellet die from abrasive samples
Shallow, thin-walled aluminum cups form the bottom and sides of the finished pellet to help reinforce XRF pellets.
$36.80 - $432.75USD / Pkg of 1000
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Description
Availability Pricing
COMPARE
33 Spec-Caps® Cups, 33 mm; 100/PK
$48.30USD / Pkg of 100
COMPARE
38 Spec-Caps® Cups, 38 mm; 100/PK
$53.60USD / Pkg of 100
COMPARE
31 Edge Flaring Tool, 31/40 mm; 1/EA
$133.50USD / Each
COMPARE
30 Spec-Caps® Cups, 30 mm; 100/PK
$47.30USD / Pkg of 100
COMPARE
33 Spec-Caps® Cups, 33 mm; 1,000/PK
$384.50USD / Pkg of 1000
COMPARE
38 Spec-Caps® Cups, 38 mm; 1,000/PK
$432.75USD / Pkg of 1000
COMPARE
31 Spec-Caps® Cups, Preflared, 31 mm; 100/PK
$36.80USD / Pkg of 100
COMPARE
31 Spec-Caps® Cups, Preflared, 31 mm; 1,000/PK
$233.25USD / Pkg of 1000
COMPARE
30 Spec-Caps® Cups, 30 mm; 1,000/PK
$384.50USD / Pkg of 1000
Showing 1- 9 of 9