Spex® VapLock™ Solvent Waste Kits for Bottles and Carboys
Collect effluent from multiple sources while minimizing harmful solvent exhaust
  • Various cap sizes available to fit your solvent container
  • 10 port manifold—1 x 1/2" NPT(F), 3 x 1/4" NPT(F), and 6 x 1/4-28 UNF(F)
Use the waste kits to minimize leaks, spills, and vapor escape from solvent waste containers. Waste kits include a 10-port manifold, exhaust filter, adapters, plugs, and tubing—everything you need to install on a typical HPLC instrument. Manifold allows solvent waste to be collected from multiple effluent streams. Exhaust filter minimize exposure to harmful chemical vapors. Fittings and tubing provide leak-tight connections minimizing spills and leaks.
$303.50 - $502.50USD / Each
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material
Availability Pricing
COMPARE
GL38 Polypropylene
$350.50USD / Each
COMPARE
38-430 PTFE
$411.50USD / Each
COMPARE
38 mm Wako Polypropylene
$307.25USD / Each
COMPARE
38 mm Wako PTFE
$368.50USD / Each
COMPARE
GL45 Polypropylene
$363.00USD / Each
COMPARE
GL45 PTFE
$419.75USD / Each
COMPARE
DIN 50 Polypropylene
$307.50USD / Each
COMPARE
51 mm PTFE
$387.25USD / Each
COMPARE
53B Polypropylene
$402.50USD / Each
COMPARE
DIN 60 Polypropylene
$325.50USD / Each
COMPARE
DIN 60 PTFE
$387.25USD / Each
COMPARE
61 mm PTFE
$368.50USD / Each
COMPARE
63 mm Polypropylene
$348.25USD / Each
COMPARE
63 mm PTFE
$398.25USD / Each
COMPARE
70 mm Nalgene Polypropylene
$405.00USD / Each
COMPARE
70 mm Nalgene PTFE
$502.00USD / Each
COMPARE
GL80 PTFE
$502.50USD / Each
COMPARE
83B Polypropylene
$371.50USD / Each
COMPARE
63 mm Fort-Pak container Polypropylene
$404.00USD / Each
COMPARE
63 mm Fort-Pak container PTFE
$485.25USD / Each
Showing 1 - 20 of 29