This product is now discontinued, View Related Products
Spex® Vaplock™ Stopcocks
Easily restrict or stop the flow of fluids
  • Clear polycarbonate (PC) body
  • Durable polyethylene (PE) handle on top of stopcock
  • Ideal for use in a variety of industries
The one-way stopcock connector involves a capped threaded female luer to fit into a male slip luer, while the three-way consists of two threaded female luers to fit into a male slip luer.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connection Type-Port 1 Connection Type-Port 2 Number of Ports
Availability Pricing
Spex VapLock™ One-Way Stopcock Conector, Female Luer to Male Luer EW-12023-27 Mfr # FCFM-001
COMPARE
Female Luer Male Luer 2
Call For Price
COMPARE
Female Luer Female Luer 3