Spilfyter Hazmat SM Sorbent Pads and Rolls
Coverstock uses spunbond material for excellent durability and virtually no linting
Hazmat sorbents are specifically designed to absorb aggressive fluids such as acids and bases. Ideal for laboratory use.
$55.50 - $382.00USD / Bag of 1
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Absorption Description
Availability Pricing
NPS Spilfyter Hazmat Yellow Spill Pads, Heavy-Weight, 16" x 18", 100/Case EW-86423-69 Mfr # S2-75
COMPARE
30 gal. (114 L) Hazmat Yellow Spill Pads, Heavy-Weight, 16" x 18", 100/Case
$144.00USD / Case of 100
NPS Spilfyter Hazmat Yellow Spill Pads, Light-Weight, 16" x 18", 200/Case EW-86423-70 Mfr # S2-70
COMPARE
44 gal. (167 L) Hazmat Yellow Spill Pads, Light-Weight, 16" x 18", 200/Case
$157.00USD / Case of 200
NPS Spilfyter Hazmat Yellow Spill Pads, Light-Weight, 12" x 12", 100/Case EW-86423-71 Mfr # S2-71
COMPARE
11 gal. (42 L) Hazmat Yellow Spill Pads, Light-Weight, 12" x 12", 100/Case
$55.50USD / Case of 100
NPS Spilfyter Hazmat Yellow Drum Top Spill Pad, Heavy-Weight, 22" diameter, 25/Case EW-86423-72 Mfr # S2-76
COMPARE
14 gal. (53 L) Hazmat Yellow Drum Top Spill Pad, Heavy-Weight, 22" diameter, 25/Case
$159.00USD / Case of 25
NPS Spilfyter Hazmat Yellow Spill Pad Roll, Heavy-Weight, 32" x 150', 1/Bag EW-86423-73 Mfr # S2-90
COMPARE
60 gal. (227 L) Hazmat Yellow Spill Pad Roll, Heavy-Weight, 32" x 150', 1/Bag
$382.00USD / Bag of 1
NPS Spilfyter Hazmat Yellow Spill Pad Roll, Heavy-Weight, 16" x 150', 2/Bag EW-86423-74 Mfr # S2-91
COMPARE
60 gal. (227 L) Hazmat Yellow Spill Pad Roll, Heavy-Weight, 16" x 150', 2/Bag
$290.00USD / Bag of 2
NPS Spilfyter Hazmat Yellow Spill Pad Roll, Light-Weight, 32" x 300', 1/Bag EW-86423-75 Mfr # S2-95
COMPARE
71 gal. (269 L) Hazmat Yellow Spill Pad Roll, Light-Weight, 32" x 300', 1/Bag
$348.00USD / Bag of 1
NPS Spilfyter Hazmat Yellow Spill Pad Roll, Light-Weight, 16" x 300', 2/Bag EW-86423-76 Mfr # S2-96
COMPARE
71 gal. (269 L) Hazmat Yellow Spill Pad Roll, Light-Weight, 16" x 300', 2/Bag
$348.00USD / Bag of 2
Showing 1- 8 of 8
Need Help with this product?
Call Us At1-847-549-7600
Live chat