Spilfyter Kolor-Safe Base Neutralizers
Three-color change lets you know when spill is completely neutralized
Protect your workers and facility from base spills by quickly and efficiently neutralizing them. Simply apply the neutralizer until the spill goes through the three-color change. Base neutralizer changes from red to blue to yellow to indicate total neutralization.
$26.00 - $61.50USD / Quart of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
NPS Spilfyter Kolor-Safe Base Neutralizer, 1 quart EW-86423-12 Mfr # 430001
COMPARE
Base Neutralizer, 1 quart
$26.00USD / Quart of 1
NPS Spilfyter Kolor-Safe Base Neutralizer, 1 gallon EW-86423-13 Mfr # 430004
COMPARE
Base Neutralizer, 1 gallon
$61.50USD / Gallon of 1
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat