Spilfyter Premium Oil-Only Sorbent Rolls
Premium rolls feature excellent absorption, strength, and low-linting
Oil-Only sorbents absorb all oil-based liquids and repel water. Ideal for use with lubricants, fuels, mineral oils, and vegetable oils. Due to low-linting and high strength, premium sorbents are designed for extended-use applications.
$244.00 - $312.75USD / Bag of 2
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Absorption Description
Availability Pricing
NPS Spilfyter Premium Oil-Only Blue Spill Pad Roll, Heavy-Weight, 16" x 150', 2/Bag EW-86423-90 Mfr # M-91
COMPARE
66 gal. (250 L) Premium Oil-Only Blue Spill Pad Roll, Heavy-Weight, 16" x 150', 2/Bag
$312.75USD / Bag of 2
NPS Spilfyter Premium Oil-Only Blue Spill Pad Roll, Heavy-Weight, 24" x 150', 1/Bag EW-86423-91 Mfr # M-92
COMPARE
50 gal. (189 L) Premium Oil-Only Blue Spill Pad Roll, Heavy-Weight, 24" x 150', 1/Bag
$244.00USD / Bag of 1
NPS Spilfyter Premium Oil-Only Blue Spill Pad Roll, Light-Weight, 16" x 300', 2/Bag EW-86423-95 Mfr # M-96
COMPARE
66 gal. (250 L) Premium Oil-Only Blue Spill Pad Roll, Light-Weight, 16" x 300', 2/Bag
$310.50USD / Bag of 2