Spilfyter Premium Yellow Hazmat Spill Socks
Surround spills or machinery to keep work areas safe
Hazmat sorbent socks are specifically designed to absorb aggressive fluids such as acids and bases. Nylon skin resists tearing while the meltblown polypropylene filler wicks liquids from surfaces for optimum absorbency.
$111.75 - $138.25USD / Case of 15
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Absorption Length (in) Width (in)
Availability Pricing
NPS Spilfyter Premium Hazmat Yellow Sock, 3" x 4', 15/Case EW-86424-25 Mfr # S2-30
COMPARE
22 gal. (84 L) 48 3
$138.25USD / Case of 15
NPS Spilfyter Premium Hazmat Yellow Sock, 3" x 10', 6/Case EW-86424-27 Mfr # S2-37
COMPARE
22 gal. (84 L) 120 3
$111.75USD / Case of 6