Spirax Sarco Cast Iron Inverted Bucket Steam Traps
  • Keep steam space free of condensate to provide maximum system efficiency
  • All stainless steel working parts provide good resistance to corrosion and wear
  • Inverted bucket steam traps rely on the difference in density between steam and condensate to drain condensate as it forms.
  • Condensate is discharged close to steam temperature
Typical applications:
  • Steam main drip stations
  • Laundry equipment
  • Industrial dryers
  • Storage tanks
$204.40 - $259.20USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Max Pressure (PSI) Outlet Port
Availability Pricing
Spirax Sarco B1H-15 1/2" 64147  Cast iron inverted bucket steam trap; 1/2" NPT(F), 15 psig EW-07600-54 Mfr # B1H-15 1/2" 64147
COMPARE
1/2" NPT(F) 15 1/2" NPT(F)
$259.20USD / Each
Spirax Sarco B1H-30 1/2" 64148  Cast iron inverted bucket steam trap; 1/2" NPT(F), 30 psig EW-07600-56 Mfr # B1H-30 1/2" 64148
COMPARE
1/2" NPT(F) 30 1/2" NPT(F)
$213.70USD / Each
Spirax Sarco B1H-75 1/2" 64149  Cast iron inverted bucket steam trap; 1/2" NPT(F), 75 psig EW-07600-58 Mfr # B1H-75 1/2" 64149
COMPARE
1/2" NPT(F) 75 1/2" NPT(F)
$204.40USD / Each
Spirax Sarco B1H-125 1/2" 64150  Cast iron inverted bucket steam trap; 1/2" NPT(F), 125 psig EW-07600-60 Mfr # B1H-125 1/2" 64150
COMPARE
1/2" NPT(F) 125 1/2" NPT(F)
$233.20USD / Each
Spirax Sarco B1H-15 3/4" 64159  Cast iron inverted bucket steam trap; 3/4" NPT(F), 15 psig EW-07600-62 Mfr # B1H-15 3/4" 64159
COMPARE
3/4" NPT(F) 15 3/4" NPT(F)
$238.30USD / Each
Spirax Sarco B1H-30 3/4" 64160  Cast iron inverted bucket steam trap; 3/4" NPT(F), 30 psig EW-07600-64 Mfr # B1H-30 3/4" 64160
COMPARE
3/4" NPT(F) 30 3/4" NPT(F)
$231.10USD / Each
Spirax Sarco B1H-75 3/4" 64161  Cast iron inverted bucket steam trap; 3/4" NPT(F), 75 psig EW-07600-66 Mfr # B1H-75 3/4" 64161
COMPARE
3/4" NPT(F) 75 3/4" NPT(F)
$216.80USD / Each
Spirax Sarco B1H-125 3/4" 64162  Cast iron inverted bucket steam trap; 3/4" NPT(F), 125 psig
Always in Stock
EW-07600-68 Mfr # B1H-125 3/4" 64162
COMPARE
3/4" NPT(F) 125 3/4" NPT(F)
$227.80USD / Each
Showing 1- 8 of 8