Spun Polypropylene Progressive-Density Cartridge 1393
Spun gradient-density construction provides greater dirt holding capacity
Ideal for removing a broad range of deformable and nondeformable particles such as sand, silt, sludge, rust, and scale. Polypropylene construction is resistant to chemical and bacterial attack. Suitable to 145°F (63°C). Note: For optimal performance, change cartridge at 25 psid.
$7.00 - $21.75USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Flow Rate Micron Rating
Availability Pricing
COMPARE
10 5 GPM at 0.6 psid 1
$7.00USD / Each
COMPARE
10 5 GPM at 0.2 psid 5
$21.75USD / Pkg of 2
COMPARE
20 10 GPM at 0.6 psid 1
$13.60USD / Each
COMPARE
20 10 GPM at 0.6 psid 5
$14.90USD / Each
COMPARE
20 5 GPM 25
$19.40USD / Each