Stainless Steel Compression T-connectors
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC).
$38.00 - $114.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
316 Stainless Steel
$41.00USD / Each
COMPARE
316 Stainless Steel
$38.00USD / Each
COMPARE
316 Stainless Steel
$49.00USD / Each
COMPARE
316 Stainless Steel
$76.00USD / Each
COMPARE
316 Stainless Steel
$114.00USD / Each