Idex® High-Pressure Stainless Steel Nuts
Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar)!
  • Constructed of durable stainless steel
These 316 stainless steel nuts (and ferrules) are rated to 20,000 psi (1380 bar) when tightened using a wrench. Recommended torque for tightening these fittings is 20 in-lbs. (2.25 N-m).
$6.20 - $150.25USD / Pkg of 10
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 1) (in) Description
Availability Pricing
Idex 316 Stainless Steel Nut, Short, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-01939-80 Mfr # U-400X
COMPARE
1/16 10-32 coned 316 Stainless Steel Nut, Short, 1/16\" OD Tubing, 10-32 Coned; 10/PK
$63.10USD / Pkg of 10
Idex 316 Stainless Steel Nut, Valco/VICI Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-02014-68 Mfr # U-320X
COMPARE
1/16 10-32 coned 316 Stainless Steel Nut, Valco/VICI Compatible, 1/16\" OD Tubing, 10-32 Coned; 10/PK
$55.60USD / Pkg of 10
Idex 316 Stainless Steel Nut, Waters Compatible, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-02014-74 Mfr # U-410X
COMPARE
1/16 10-32 coned 316 Stainless Steel Nut, Waters Compatible, 1/16\" OD Tubing, 10-32 Coned; 10/PK
$62.80USD / Pkg of 10
Idex 316 Stainless Steel Nut, 1/8" OD Tubing, 1/4-28 Coned; 1/EA EW-02021-07 Mfr # C-235
COMPARE
1/8 1/4-28 coned 316 Stainless Steel Nut, 1/8\" OD Tubing, 1/4-28 Coned; 1/EA
$13.80USD / Each
Idex 316 Stainless Steel Nut, 1/8" OD Tubing, 1/4-28 Coned; 10/PK EW-02021-08 Mfr # C-235X
COMPARE
1/8 1/4-28 coned 316 Stainless Steel Nut, 1/8\" OD Tubing, 1/4-28 Coned; 10/PK
$128.00USD / Pkg of 10
Idex 316 Stainless Steel Nut, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-02022-32 Mfr # 6000-263
COMPARE
1/16 10-32 coned 316 Stainless Steel Nut, 1/16\" OD Tubing, 10-32 Coned; 10/PK
$64.70USD / Pkg of 10
Idex 316 Stainless Steel Nut, Extra Long, 1/16" OD Tubing, 10-32 Coned; 10/PK EW-02022-34 Mfr # 6000-265
COMPARE
1/16 10-32 coned 316 Stainless Steel Nut, Extra Long, 1/16\" OD Tubing, 10-32 Coned; 10/PK
$111.25USD / Pkg of 10
Idex 316 Stainless Steel Nut, Short, 1/16" OD Tubing, 10-32 Coned; 1/EA EW-02022-36 Mfr # U-400
COMPARE
1/16 10-32 coned 316 Stainless Steel Nut, Short, 1/16\" OD Tubing, 10-32 Coned; 1/EA
$6.20USD / Each
Idex 316 Stainless Steel Nut, Waters Compatible, 1/16" OD Tubing, 10-32 Coned; 1/EA EW-02022-38 Mfr # U-410
COMPARE
1/16 10-32 coned 316 Stainless Steel Nut, Waters Compatible, 1/16\" OD Tubing, 10-32 Coned; 1/EA
$6.80USD / Each
Idex 316 Stainless Steel Nut, 1/16" OD Tubing, M6 Coned; 1/EA EW-02022-39 Mfr # U-450
COMPARE
1/16 M6 coned 316 Stainless Steel Nut, 1/16\" OD Tubing, M6 Coned; 1/EA
$16.40USD / Each
Idex 316 Stainless Steel Nut, 1/16" OD Tubing, M6 Coned; 10/PK EW-02022-40 Mfr # U-450X
COMPARE
1/16 M6 coned 316 Stainless Steel Nut, 1/16\" OD Tubing, M6 Coned; 10/PK
$150.25USD / Pkg of 10
Showing 1- 11 of 11