Stainless Steel Multiple Filter Housings
Stack up to 20 double open end cartridges in one housing!
Clearly labeled ports—easy to install and maintain
Stainless steel housings are great for drinking water applications in schools, restaurants, institutions, and industrial plants. Housings hold from 4 to 20 double open end cartridges--filter a large volume of water every minute! Housings feature durable 304 stainless steel construction with epoxy finish to provide better chemical compatibility. All ports are clearly labeled for easy installation.
Maximum pressure is 125 psi; maximum temperature is 300°F (149<>degC). Clean and dirty drains are 1/4", inlet and outlet ports are 2" NPT (M). Order cartridges separately. Buna N seal.
$1,685.50 - $1,926.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Number of Cartridges
Availability Pricing
Pentair 156025-02 304 stainless steel housing for 4 filters EW-29810-05 Mfr # 156025-02
COMPARE
4
$1,685.50USD / Each
Pentair 156026-02 304 stainless steel housing for 8 filters EW-29810-10 Mfr # 156026-02
COMPARE
8
$1,719.00USD / Each
Pentair 156027-02 304 stainless steel housing for 12 filters EW-29810-15 Mfr # 156027-02
COMPARE
12
$1,926.00USD / Each