Washable Static Control Lab Coats
Made from 94% polyester and 6% carbon-suffused nylon – excellent control of static electricity
  • Adjustable sleeves feature a snap at each wrist for easy grounding
Material maintains static resistance even after multiple washings. Coats close easily with snap closures. Adjustable sleeves feature a grounding snap at each wrist. Each coat has two lower pockets and one left breast pocket.
Compliance: ESD STM2.1-1997.
$64.00 - $93.90USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Size
Availability Pricing
VF Workwear Static Control/Antistatic Lab Coat, 33" L, Small EW-86297-40 Mfr # KK26BL-SMALL (PACK BY PERSON)
COMPARE
Blue 33 Small
$93.90USD / Each
VF Workwear Static Control/Antistatic Lab Coat, 33" L, Medium EW-86297-41 Mfr # KK26BL-MEDIUM (PACK BY PERSON)
COMPARE
Blue 33 Medium
$64.00USD / Each
VF Workwear Static Control/Antistatic Lab Coat, 33" L, Large EW-86297-42 Mfr # KK26BL-LARGE (PACK BY PERSON)
COMPARE
Blue 33 Large
$64.00USD / Each
VF Workwear Static Control/Antistatic Lab Coat, 33" L, X-Large EW-86297-43 Mfr # KK26BL-XLARGE (PACK BY PERSON)
COMPARE
Blue 33 X-large
$67.10USD / Each
VF Workwear Static Control/Antistatic Lab Coat, 41.5" L, Small EW-86297-44 Mfr # KK28BL-SMALL (PACK BY PERSON)
COMPARE
Blue 41.5 Small
$66.60USD / Each
VF Workwear Static Control Lab Coats, medium, 41.5"L EW-86297-45 Mfr # KK28BL-MEDIUM (PACK BY PERSON)
COMPARE
Blue 41.5 Medium
$67.00USD / Each
VF Workwear Static Control/Antistatic Lab Coat, 41.5" L, Large EW-86297-46 Mfr # KK28BL-LARGE (PACK BY PERSON)
COMPARE
Blue 41.5 Large
$67.00USD / Each
VF Workwear Static Control Lab Coats, X-large, 41.5"L EW-86297-47 Mfr # KK28BL-XLARGE (PACK BY PERSON)
COMPARE
Blue 41.5 X-large
$67.50USD / Each
Showing 1- 8 of 8