Cole-Parmer® Essentials Sterile Petri Dishes
Sterilized dishes are ready to use
  • Constructed clear polystyrene
  • Lids contain stacking rings for increased stability
  • Ventilation ribs reduce condensation and allow air circulation
Disposable polystyrene dishes have convenient stacking ridges for enhanced stability. Consistent flatness allows for even media distribution. All dishes are supplied sterile (gamma-irradiated) and come packaged 20 dishes/bag.
$223.50 - $225.00USD / Box of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Outside Diameter (OD) (cm) Description
Availability Pricing
COMPARE
Polystyrene 10 Sterile Petri Dishes; 100 mm dia x 15 mm H; 500/box
$223.50USD / Box of 500
COMPARE
Polystyrene 6 Sterile Petri Dishes; 60 mm dia x 15 mm H; 500/box
$225.00USD / Box of 500