Sterile Sampling Spoons, Polystyrene (PS)
Individually sterilized and bagged
  • Long handle reaches easily into jars, bottles, and other confined areas; minimizes the potential of personal contact with sampled material
  • FDA grade materials; safe for food, drug, and cosmetic contact
Designed to gather small amounts of sample material, these sterile polystyrene plastic sampling spoons are available in four bowl sizes. All are individually packaged, sterilized by gamma irradiation, lot stamped for accurate tracing, and shipped with a Certificate of Processing for Sterilization.
$190.00 - $239.00USD / Case of 200
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Sterile
Availability Pricing
Sterile polystyrene sampling spoon, 1/4 tsp EW-06263-10 Mfr # BMP-SO-400
COMPARE
1.25 Polystyrene Yes
$239.00USD / Case of 200
Sterile polystyrene sampling spoon, 1/2 tsp EW-06263-20 Mfr # BMP-SO-402
COMPARE
2.5 Polystyrene Yes
$229.00USD / Case of 200
Sterile polystyrene sampling spoon, 1 tsp
Always in Stock
EW-06263-30 Mfr # BMP-SO-404
COMPARE
5 Polystyrene Yes
$203.00USD / Case of 200
Sterile polystyrene sampling spoon, 1 tbsp
Always in Stock
EW-06263-40 Mfr # BMP-SO-406
COMPARE
14.8 Polystyrene Yes
$190.00USD / Case of 200