Sticht Compact and Full Size Vibrating Reed Tachometers
Accurate - Rugged - Dependable
  • Compact size tachometers have smaller and fewer reeds
  • Ideal for tool case or pocket use
  • Popular in the textile industry for measuring speeds of sewing machines
Because these tachometers measure speeds by contact, this type of tachometer is the solution to many speed measuring problems, especially checking speeds of totally enclosed rotating equipment. These tachometers operate on the well known and time tested principle of resonance. The instrument is simply held against motor, turbine, pump, compressor, or other rotating equipment and the speed is shown by the vibration of a steel reed which is tuned to a certain standard speed. Ruggedly constructed, permanent accuracy, simple to use. The instrument imposes no load and cannot be damaged by overspeeding. Accuracy is 0.5% of rated speed. Other typical applications are outboard motors, electric shavers, centrifuges, etc.
$434.75 - $1,299.00USD / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max RPM Range (rpm) Description
Availability Pricing
Sticht 9275 Compact Vibrating Reed Tachometer; 1000 to 4000 rpm EW-98805-97 Mfr # 9275
COMPARE
Analog 4000 Compact Vibrating Reed Tachometer; 1000 to 4000 rpm
$659.00USD / Each
Sticht 9281 Compact Vibrating Reed Tachometer; 2500 to 6500 rpm EW-98805-98 Mfr # 9281
COMPARE
Analog 6500 Compact Vibrating Reed Tachometer; 2500 to 6500 rpm
$575.50USD / Each
Sticht 9284 Compact Vibrating Reed Tachometer; 3500 to 7500 rpm EW-98806-00 Mfr # 9284
COMPARE
Analog 7500 Compact Vibrating Reed Tachometer; 3500 to 7500 rpm
$434.75USD / Each
Sticht 9280 Compact Vibrating Reed Tachometer; 9000 to 15,000 rpm EW-98806-02 Mfr # 9280
COMPARE
Analog 15000 Compact Vibrating Reed Tachometer; 9000 to 15,000 rpm
$557.50USD / Each
Sticht 6228C 6228-C Vibrating Reed Tachometer; 600 to 1500 rpm EW-98806-04 Mfr # 6228C
COMPARE
Analog 600 6228-C Vibrating Reed Tachometer; 600 to 1500 rpm
$999.50USD / Each
Sticht 6235 Vibrating Tachometer 1750/2500 2500/4000 Range Rpm EW-98806-06 Mfr # 6235
COMPARE
Analog 4000 Vibrating Tachometer 1750/2500 2500/4000 Range Rpm
$701.50USD / Each
Sticht 6239 Vibrating Reed Tachometer; 1000 to 5500 rpm EW-98806-07 Mfr # 6239
COMPARE
Analog 5500 Vibrating Reed Tachometer; 1000 to 5500 rpm
$835.50USD / Each
Sticht 6246 Vibrating Reed Tachometer; 3000 to 7500 rpm EW-98806-08 Mfr # 6246
COMPARE
Analog 7500 Vibrating Reed Tachometer; 3000 to 7500 rpm
$663.00USD / Each
Sticht 6248 Vibrating Reed Tachometer; 4500 to 9000 rpm EW-98806-09 Mfr # 6248
COMPARE
Analog 9000 Vibrating Reed Tachometer; 4500 to 9000 rpm
$663.00USD / Each
Sticht 6250 Vibrating Reed Tachometer; 6000 to 12,000 rpm EW-98806-10 Mfr # 6250
COMPARE
Analog 12000 Vibrating Reed Tachometer; 6000 to 12,000 rpm
$882.50USD / Each
Sticht 6355 Vibrating Reed Tachometer; 1000 to 6000 rpm EW-98806-13 Mfr # 6355
COMPARE
Analog 6000 Vibrating Reed Tachometer; 1000 to 6000 rpm
$1,173.00USD / Each
Sticht 6359 Vibrating Reed Tachometer; 600 to 8000 rpm EW-98806-14 Mfr # 6359
COMPARE
Analog 8000 Vibrating Reed Tachometer; 600 to 8000 rpm
$1,299.00USD / Each
Sticht 6360 Vibrating Reed Tachometer; 3000 to 12,000 rpm EW-98806-15 Mfr # 6360
COMPARE
Analog 3000 Vibrating Reed Tachometer; 3000 to 12,000 rpm
$1,172.00USD / Each
Sticht 6357 Vibrating Reed Tachometer; 6000 to 24,000 rpm EW-98806-16 Mfr # 6357
COMPARE
Analog 24000 Vibrating Reed Tachometer; 6000 to 24,000 rpm
$1,039.00USD / Each
Sticht 6358 Vibrating Reed Tachometer; 7500 to 30,500 rpm EW-98806-17 Mfr # 6358
COMPARE
Analog 7500 Vibrating Reed Tachometer; 7500 to 30,500 rpm
$1,238.00USD / Each
Sticht 9283 Compact Vibrating Reed Tachometer; 1000 to 6000 rpm EW-98806-23 Mfr # 9283
COMPARE
Analog 1500 Compact Vibrating Reed Tachometer; 1000 to 6000 rpm
$547.00USD / Each
Sticht 6356 Vibrating Reed Tachometer; 1500 to 7500 rpm EW-98806-24 Mfr # 6356
COMPARE
Analog 3500 Vibrating Reed Tachometer; 1500 to 7500 rpm
$944.00USD / Each
Showing 1- 17 of 17