Stuart Heating Blocks
  • Interchangeable heating blocks accommodate various tubes and plates and offer excellent thermal conductivity
  • Block heaters sold separately
All blocks are made of anodized aluminum (except 36610-43). Most blocks feature a thermometer hole to let you easily monitor block temperature. Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Flexi-blocks use autoclavable high-density ceramic beads and are available in two sizes—double or triple width. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.
$164.00 - $1,710.00USD / Each
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Block Heater Insert, triple wide, Flexi-block
$574.00USD / Each
COMPARE
Block Heater Insert, solid stainless steel
$185.00USD / Each
COMPARE
Block Heater Insert, 10 mm diameter holes
$184.00USD / Each
COMPARE
Block Heater Insert, 12 mm dia x 47 mm deep
$184.00USD / Each
COMPARE
Block Heater Insert, 12 mm dia x 33 mm deep
$195.00USD / Each
COMPARE
Block Heater Insert, 12 mm diameter holes
$167.00USD / Each
COMPARE
Block Heater Insert, 13 mm diameter holes
$184.00USD / Each
COMPARE
Block Heater Insert, 16 mm diameter holes
$183.00USD / Each
COMPARE
Block Heater Insert, 17 mm diameter holes
$178.00USD / Each
COMPARE
Block Heater Insert, 19 mm diameter holes
$184.00USD / Each
COMPARE
Block Heater Insert, cuvette and pipette channels
$187.00USD / Each
COMPARE
Block Heater Insert, 20 x 1.5 mL tubes
$183.00USD / Each
COMPARE
Block Heater Insert, 25 mm diameter holes
$184.00USD / Each
COMPARE
Block Heater Insert, 28 mm diameter holes
$184.00USD / Each
COMPARE
Block Heater Insert, 30 x 0.5 mL tubes
$224.00USD / Each
COMPARE
Block Heater Insert, 30 mm diameter holes
$187.00USD / Each
COMPARE
Block Heater Insert, 33 mm diameter holes
$184.00USD / Each
COMPARE
Block Heater Insert, 384 well plate
$1,710.00USD / Each
COMPARE
Block Heater Insert, 48 x 0.2 mL tubes
$222.00USD / Each
COMPARE
Block Heater Insert, 96 well plate
$607.00USD / Each
Showing 1 - 20 of 25
Need Help with this product?
Call Us At1-847-549-7600
Live chat