Stuart Orbital Shakers
Maintain consistent shaking speeds, even over long periods of time
  • Smooth orbital shaking action with 16-mm orbit — ideal for larger samples and multiwell plates
  • Built-in digital timer and variable speed control to 300 rpm
  • Included rubber mat helps hold short-flat vessels securely in place
These orbital shaker provides a smooth, uniform circular motion with a 16-mm orbit. Speed and time may be varied from 30 to 300 rpm, and 1 to 999 minutes — easily set and read on the digital display. The shaking action is ideal for samples of 0.5 to 5 mL in multiwell plates, dishes, and petri dishes. Use in incubators, and environmental or cold chambers up to 104°F (40°C) and 80% humidity.
$1,130.00 - $1,810.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Platform Length (in) Platform Width (in) Power (VAC)
Availability Pricing
COMPARE
8.625 8.625 120
$1,160.00USD / Each
COMPARE
8.625 8.625 230
$1,130.00USD / Each
COMPARE
13.1875 13.1875 120
$1,810.00USD / Each
COMPARE
13.1875 13.1875 230
$1,740.00USD / Each
Showing 1- 4 of 4
Need Help with this product?
Call Us At1-847-549-7600
Live chat