Cole-Parmer Stuart® Shaking Incubators
Shaker and incubator combination makes these ideal for cell culture procedures
  • Microprocessor controlled shaking speed ensures accuracy
  • Forced air circulation ensures uniform temperature inside the chamber
  • Easily set and view temperature and speed on the digital display
  • Unique retractable platform offers easy loading and unloading
These shaking incubators provide an orbital shaking motion to ensure even mixing while promoting bacterial growth. All models feature a timer which can be set from 1 second to 9 days. Once the timer has counted down, an alarm will sound and the shaking action will stop while the incubator will continue to run. The retractable platform allows easy access to the samples at the back of the incubator yet securely locks in place during operation. Built-in USB connection and dedicated software allow long-term monitoring of the incubator temperature via a PC. Order models with IQ/OQ (Installation Qualification/Operational Qualification) documentation to help you comply with regulatory agency requirements.
$3,140.00 - $8,515.00USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Operating Temperature (° C) Max Speed (rpm) Power (VAC)
Availability Pricing
COMPARE
60 300 120
$3,865.00USD / Each
COMPARE
60 300 220
$4,535.00USD / Each
COMPARE
60 300 230
$3,920.00USD / Each
COMPARE
60 300 230
$5,545.00USD / Each
COMPARE
60 1250 120
$3,720.00USD / Each
COMPARE
60 1250 230
$3,140.00USD / Each
COMPARE
60 1250 230
$3,585.00USD / Each
COMPARE
60 300 120
$6,425.00USD / Each
COMPARE
60 300 230
$6,035.00USD / Each
COMPARE
60 300 230
$6,720.00USD / Each
COMPARE
60 300 120
$8,515.00USD / Each
COMPARE
60 300 230
$8,515.00USD / Each
COMPARE
60 300 230
$7,645.00USD / Each
Showing 1- 13 of 13