Cole-Parmer® WA-200 Series Wrist-Action Shaker
Eliminate wrist strain
  • Vigorous mixing action simulates hand shaking
  • Ideal for extractions
The variable-speed shaker features two side arms with adjustable clamps capable of holding eight flasks or bottles up to 500 mL capacity. Use for prolonged continuous shaking or set the built-in analog timer from 10 to 60 minutes and unit automatically turns off when done. Electronic feedback controller ensures a constant speed, irrespective of load.
$1,954.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (Hz) Power (VAC)
Availability Pricing
COMPARE
50 230
$1,954.00USD / Each