Sub-Micro Quartz Cuvette for Fluorometry
Ideal for fluorometry applications
  • Constructed of quartz
Available for fluorimetry (three polished windows). Match the cuvette center height to instrument light beam height; see individual cuvette specification table (either 8.5 or 15 mm) to verify correct manufacturer beam height.
$1,246.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Beam Height Zd (mm) Max Capacity (MicroL) Pathlength (mm)
Availability Pricing
COMPARE
8.5 160 10
$1,246.00USD / Each