This product is now discontinued, View Related Products
Submersible 3-ft pH Electrodes
Won’t fail even in tough industrial environments
  • Excellent for measuring large industrial tanks and processes
  • Permanently potted in sturdy ABS plastic housing
  • Rugged double-junction designs
  • Three-foot probe length for multiple applications
Choose from a variety of unique electrode designs for use in tough industrial environments. All models feature BNC pH connectors to match a variety of meters and controllers. Electrodes with temperature compensation feature 100 Ω RTD temperature elements with mini-audio plug connections. These combination electrodes are permanently potted in a 3 ft L x 11⁄16 inch dia ABS plastic sheath—submersible up to the cable. These unique electrodes are ideal for measurements in open containers
such as tanks or troughs.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
ATC Element Connector Electrode Type
Availability Pricing
COMPARE
BNC Double junction
Call For Price
COMPARE
100 Ω Pt RTD BNC Double junction