Swing Sampler
Sampler swings allowing collections from a variety of different angles – including 90°
  • Easily collects samples from horizontally flowing streams
Use the swing sampler to obtain liquids, powders, or even small solids. Includes one 960-mL bottle and fits almost any other laboratory sample bottles.
$194.00 - $248.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Bottle Type Length (ft) Sample Volume
Availability Pricing
Nasco Sampling B01310WA Swing Sampler, polyethylene & polyurethane, 6 to 12 ft depth
Always in Stock
EW-99155-30 Mfr # B01310WA
COMPARE
Polyethylene 12 960 mL
$194.00USD / Each
Nasco Sampling B01366WA Swing Sampler, polyethylene & polyurethane, 8 to 24 ft depth
Always in Stock
EW-99155-32 Mfr # B01366WA
COMPARE
Polyethylene 24 960 mL
$248.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat