Tapered Tissue Grinders
Improve grinding efficiency to save time
  • Pestle is compatible with overhead stirrers having collets or chucks for 0.25" diameter mixer shafts
This design incorporates a longer tapered surface on mortar and pestle. Grinders feature a PTFE pestle with 0.25" stainless steel rod.
$138.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
DWK Life Sciences (Wheaton) 358133 Tapered Tissue Grinder, Glass, 1 mL; 1/Cs EW-04468-40 Mfr # 358133
COMPARE
1
$138.00USD / Each