Taylor Digital Food Portion Control Scale

Monitor portion sizes quickly and easily
  • Easy-to-read LCD display
  • Auto-shutoff function helps conserve power
  • Removable stainless steel platform for easy cleaning
These compact digital portion control scales easily handle all your foodservice portioning needs. Features include low battery warning light, overload indicator, and tare function which accurately weigh products and ingredients while still in the container. Scales are available in 21 oz, 2 lb, 11 lb and 22 lb capacities.
$65.00 - $182.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Calibration Type Capacity (lbs) Description
Availability Pricing
Taylor TE32FT Precision Digital Portion Control Scale, 2 lb x .01 oz
Always in Stock
EW-11010-14 Mfr # TE32FT
COMPARE
External 2 Precision Digital Portion Control Scale, 2 lb x .01 oz
$65.00USD / Each
Taylor TE10FT Precision Digital Portion Control Scale, 11 lb x .1 oz EW-11010-15 Mfr # TE10FT
COMPARE
External 11 Precision Digital Portion Control Scale, 11 lb x .1 oz
$70.50USD / Each
Taylor TE22FT Precision Digital Portion Control Scale, 22 lb x .1 oz EW-11010-16 Mfr # TE22FT
COMPARE
External 22 Precision Digital Portion Control Scale, 22 lb x .1 oz
$135.00USD / Each
Taylor TE21P Precision Digital Portion Control Scale, 21 oz. EW-11010-17 Mfr # TE21P
COMPARE
External 1.3 Precision Digital Portion Control Scale, 21 oz.
$182.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat