Cole-Parmer® IC-300 Series High-Stability Digital Immersion Circulator
Offers wide temperature range of -40 to 200°C
  • High ±0.005°C temperature stability
  • Bidirectional RS-232 capability
This versatile immersion circulator can be used in several ways. Use as a stand-alone water bath temperature controller, a pump for recirculating chilled liquid from an external chiller, or a pump for providing temperature control to an external device. Immersion circulator accurately heats up to 200°C and safely controls the temperature of liquid in the bath while maintaining excellent temperature stability. A bidirectional RS-232 interface lets you control bath remotely and download data to a computer for further analysis.
A digital display, overtemperature indicator light, pressure pump, and low-liquid level cutoff make the circulator easy and safe to use. Reach below ambient temperatures by using the cooling coil 12122-87 connected to an optional external chiller.
$2,125.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
200 115 High-Stability Digital Immersion Circulator; 120 VAC, 50/60 Hz
$2,125.00USD / Each
COMPARE
200 230 High-Stability Digital Immersion Circulator; 230 VAC, 50/60 Hz
$2,125.00USD / Each