Techne Plant and Animal Allergen and Speciation Detection Kits for qPCR
Quickly and accurately verify the allergens and speciation in the food source
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Great value at 100 tests per kit
Use the allergen and speciation kits to clarify sources and types of products where accurate results are critical. Order a complete kit with mastermix or select a kit without mastermix and use your own.
$978.00 - $1,224.00USD / Each
66 variations of this product are available.
See All Product Options Below
66 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mastermix Included Test Type Description
Availability Pricing
COMPARE
No DNA Specific Detection Kit for Apium graveolens var. dulce (celery) and A. graveolens var. rapaceum (celeriac), without mastermix
$978.00USD / Each
COMPARE
No DNA Specific Detection Kit for Bubalus bubalis (water buffalo), without mastermix
$978.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for Bertholletia excelsa (Brazil nut), with mastermix
$1,224.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for Capra hircus (goat), with mastermix
$1,224.00USD / Each
COMPARE
No DNA Specific Detection Kit for Carya illinoinensis (pecan), without mastermix
$978.00USD / Each
COMPARE
No DNA Specific Detection Kit for Gadus morhua (cod), without mastermix
$978.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for Gadus morhua (cod), with mastermix
$1,224.00USD / Each
COMPARE
No DNA Specific Detection Kit for Juglans regia (walnut), without mastermix
$978.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for Melanogrammus aeglefinus (haddock), with mastermix
$1,224.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for Macadamia integrifolia (Macadamia), with mastermix
$1,224.00USD / Each
COMPARE
No DNA Specific Detection Kit for Mus musculus (mouse), without mastermix
$978.00USD / Each
COMPARE
No DNA Specific Detection Kit for Ovis aries (sheep), without mastermix
$978.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for Ovis aries (sheep), with mastermix
$1,224.00USD / Each
COMPARE
No DNA Specific Detection Kit for Prunus dulcis (almond), without mastermix
$978.00USD / Each
COMPARE
No DNA Specific Detection Kit for Pleuronectes platessa (European plaice), without mastermix
$978.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for Pleuronectes platessa (European plaice), with mastermix
$1,224.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for Pistacia vera (pistacio), with mastermix
$1,224.00USD / Each
COMPARE
No DNA Specific Detection Kit for Struthio camelus (ostrich), without mastermix
$978.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for universal fish detection, with mastermix
$1,224.00USD / Each
COMPARE
Yes DNA Specific Detection Kit for universal meat detection, with mastermix
$1,224.00USD / Each
Showing 1 - 20 of 66