Techne® Sample Concentrators
Quickly concentrate multiple samples in a block at once
  • Variable needle position suits various Dri-Block® inserts
  • Needles, block, and block heater sold separately
Concentrator blows an inert gas over the surface of your samples via stainless steel needles (not included). Blown gas in combination with the heat from the heater produce ideal conditions for fast efficient evaporation. Insert needles into a silicone membrane in any configuration. Fully adjustable gas reservoir provides accurate height control. Maximum vertical traveling distance is 32 cm. Maximum gas usage is 15 L/min. Order PTFE-coated needles for use with corrosive samples.
$27.00 - $1,390.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Sample Concentrator for Tubes
$1,390.00USD / Each
COMPARE
Sample Concentrator for 96-Well Plates
$1,390.00USD / Each
COMPARE
Sample Concentrator Needles, SS, 76 mm; 100/Pk
$454.00USD / Each
COMPARE
Sample Concentrator Needles, SS, 127 mm; 100/Pk
$574.00USD / Each
COMPARE
Sample Concentrator Needles, PTFE, 127 mm; 100/Pk
$1,120.00USD / Each
COMPARE
Sample Concentrator Needles, PTFE, 76 mm; 100/Pk
$913.00USD / Each
COMPARE
$27.00USD / Each
Showing 1- 7 of 7
Need Help with this product?
Call Us At1-847-549-7600
Live chat