TEDIA® 1,2-Dichloroethane

High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$459.00 - $615.00USD / Case of 6

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Tedia 1,2-Dichloroethane, Absolv Grade; 6 x 1 L, Amber Glass EW-15000-41 Mfr # DA1420-002
COMPARE
Absolv 6000 1,2-Dichloroethane, Absolv Grade; 6 x 1 L, Amber Glass
$459.00USD / Case of 6
Tedia 1,2-Dichloroethane, HPLC/Spectrophotometric Grade; 4 x 4 L, Amber Glass EW-15000-56 Mfr # DS1421-001
COMPARE
HPLC and Spectrophotometric 16000 1,2-Dichloroethane, HPLC/Spectrophotometric Grade; 4 x 4 L, Amber Glass
$615.00USD / Case of 4
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat