This product is now discontinued, View Related Products

TEDIA® Acetone

High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.
  • Quality and consistency your lab can count on
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

11 variations of this product are available.

See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Tedia Acetone, Absolv Grade, 4 x 4 L EW-15000-00 Mfr # AA1111-001
COMPARE
Absolv 16000 Acetone, Absolv Grade, 4 x 4 L
Call For Price
Tedia Acetone, Absolv Grade; 6 x 1 L, Amber Glass EW-15000-01 Mfr # AA1111-002
COMPARE
Absolv 6000 Acetone, Absolv Grade; 6 x 1 L, Amber Glass
Call For Price
Tedia Acetone, Absolv Grade; 4 L, Amber Glass EW-15000-02 Mfr # AA1111-048
COMPARE
Absolv 4000 Acetone, Absolv Grade; 4 L, Amber Glass
Call For Price
Tedia Acetone, GC Grade, 4 x 4 L EW-15000-06 Mfr # AG2800-001
COMPARE
GC 16000 Acetone, GC Grade, 4 x 4 L
Call For Price
Tedia Acetone, GC Grade, 4 L EW-15000-07 Mfr # AG2800-048
COMPARE
GC 4000 Acetone, GC Grade, 4 L
Call For Price
Tedia Acetone, ACS Reagent Grade; 6 x 1 L, Amber Glass EW-15000-17 Mfr # AR0114-002
COMPARE
ACS Reagent 6000 Acetone, ACS Reagent Grade; 6 x 1 L, Amber Glass
Call For Price
Tedia Acetone, ACS Reagent Grade, 20 L EW-15000-18 Mfr # AR0114-007
COMPARE
ACS Reagent 20000 Acetone, ACS Reagent Grade, 20 L
Call For Price
Tedia Acetone, ACS Reagent Grade; 1 L, Amber Glass EW-15000-19 Mfr # AR0114-065
COMPARE
ACS Reagent 1000 Acetone, ACS Reagent Grade; 1 L, Amber Glass
Call For Price
Tedia Acetone, HPLC/Spectrophotometric Grade, 4 x 4 L EW-15000-21 Mfr # AS1112-001
COMPARE
HPLC and Spectrophotometric 16000 Acetone, HPLC/Spectrophotometric Grade, 4 x 4 L
Call For Price
Tedia Acetone, HPLC/Spectrophotometric Grade, 6 x 1 L EW-15000-22 Mfr # AS1112-002
COMPARE
HPLC and Spectrophotometric 6000 Acetone, HPLC/Spectrophotometric Grade, 6 x 1 L
Call For Price
Tedia Acetone, HPLC/Spectrophotometric Grade; 1 L, Amber Glass EW-15000-23 Mfr # AS1112-065
COMPARE
HPLC and Spectrophotometric 1000 Acetone, HPLC/Spectrophotometric Grade; 1 L, Amber Glass
Call For Price
Showing 1- 11 of 11

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat