TEDIA® Dichloromethane

High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.
  • Quality and consistency your lab can count on
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$321.75 - $625.50USD / Case of 6

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Tedia Dichloromethane, Absolv Grade, 4 x 4 L EW-15000-43 Mfr # DA1431-001
COMPARE
Absolv 16000 Dichloromethane, Absolv Grade, 4 x 4 L
$625.50USD / Case of 4
Tedia Dichloromethane, Absolv Grade, 6 x 1 L EW-15000-44 Mfr # DA1431-002
COMPARE
Absolv 6000 Dichloromethane, Absolv Grade, 6 x 1 L
$321.75USD / Case of 6
Tedia Dichloromethane, GC Grade, 4 x 4 L EW-15000-46 Mfr # DG2810-001
COMPARE
GC 16000 Dichloromethane, GC Grade, 4 x 4 L
$389.00USD / Case of 4
Tedia Dichloromethane, ACS Reagent Grade, 4 x 4 L EW-15000-53 Mfr # DR0440-001
COMPARE
ACS Reagent 16000 Dichloromethane, ACS Reagent Grade, 4 x 4 L
$464.00USD / Case of 4
Tedia Dichloromethane, HPLC/Spectrophotometric Grade, 4 x 4 L EW-15000-57 Mfr # DS1432-001
COMPARE
HPLC and Spectrophotometric 16000 Dichloromethane, HPLC/Spectrophotometric Grade, 4 x 4 L
$330.25USD / Case of 4
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat