TEDIA® Methyl Alcohol

High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$37.25 - $841.00USD / Each

18 variations of this product are available.

See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Form Volume (mL) Description
Availability Pricing
Tedia Methyl Alcohol, Absolv Grade; 6 x 1 L, Amber Glass EW-15001-08 Mfr # MA1921-002
COMPARE
Liquid 6000 Methyl Alcohol, Absolv Grade; 6 x 1 L, Amber Glass
$256.00USD / Case of 6
Tedia Methyl Alcohol, Absolv Grade, 1 L, Amber Glass EW-15001-09 Mfr # MA1921-065
COMPARE
Liquid 1000 Methyl Alcohol, Absolv Grade, 1 L, Amber Glass
$57.50USD / Each
Tedia Methyl Alcohol, GC Grade; 4 x 4 L, Amber Glass EW-15001-10 Mfr # MG2826-001
COMPARE
Liquid 16000 Methyl Alcohol, GC Grade; 4 x 4 L, Amber Glass
$219.00USD / Case of 4
Tedia Methyl Alcohol, GC Grade; 4 L, Amber Glass EW-15001-11 Mfr # MG2826-048
COMPARE
Liquid 4000 Methyl Alcohol, GC Grade; 4 L, Amber Glass
$101.00USD / Each
Tedia Methyl Alcohol, Purge and Trap Grade; 6 x 1 L, Amber Glass EW-15001-13 Mfr # MP1924-002
COMPARE
Liquid 6000 Methyl Alcohol, Purge and Trap Grade; 6 x 1 L, Amber Glass
$841.00USD / Case of 6
Tedia Methyl Alcohol, Histological Grade; 4 x 4 L, PVC EW-15001-14 Mfr # MR1268-012
COMPARE
Liquid 16000 Methyl Alcohol, Histological Grade; 4 x 4 L, PVC
$127.00USD / Case of 4
Tedia Methyl Alcohol, Histological Grade; 20 L, Poly Pail EW-15001-15 Mfr # MR1268-031
COMPARE
Liquid 20000 Methyl Alcohol, Histological Grade; 20 L, Poly Pail
$179.00USD / Each
Tedia Methyl Alcohol, ACS Reagent Grade; 4 x 4 L, Amber Glass EW-15001-16 Mfr # MR1272-001
COMPARE
Liquid 16000 Methyl Alcohol, ACS Reagent Grade; 4 x 4 L, Amber Glass
$151.00USD / Case of 4
Tedia Methyl Alcohol, ACS Reagent Grade; 6 x 1 L, Amber Glass EW-15001-17 Mfr # MR1272-002
COMPARE
Liquid 6000 Methyl Alcohol, ACS Reagent Grade; 6 x 1 L, Amber Glass
$118.00USD / Case of 6
Tedia Methanol, ACS Reagent Grade; 20 L, Metal Pail EW-15001-19 Mfr # MR1272-007
COMPARE
Liquid 20000 Methanol, ACS Reagent Grade; 20 L, Metal Pail
$198.00USD / Each
Tedia Methyl Alcohol, ACS Reagent Grade; 1 L, Amber Glass EW-15001-20 Mfr # MR1272-065
COMPARE
Liquid 1000 Methyl Alcohol, ACS Reagent Grade; 1 L, Amber Glass
$37.25USD / Each
Tedia methanol Low water; 4 x 4 L, Amber Glass EW-15001-21 Mfr # MR1274-001
COMPARE
Liquid 16000 methanol Low water; 4 x 4 L, Amber Glass
$175.00USD / Case of 4
Tedia Methyl Alcohol Low water; 4 L, Amber Glass EW-15001-22 Mfr # MR1274-048
COMPARE
Liquid 4000 Methyl Alcohol Low water; 4 L, Amber Glass
$69.50USD / Each
Tedia Methyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, Amber Glass EW-15001-33 Mfr # MS1922-001
COMPARE
Liquid 16000 Methyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, Amber Glass
$184.00USD / Case of 4
Tedia Methyl Alcohol, HPLC/Spectrophotometric Grade; 6 x 1 L, Amber Glass EW-15001-34 Mfr # MS1922-002
COMPARE
Liquid 6000 Methyl Alcohol, HPLC/Spectrophotometric Grade; 6 x 1 L, Amber Glass
$150.00USD / Case of 6
Tedia MS1922-006 Methyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, PVC SKU 1500135 EW-15001-35 Mfr # MS1922-006
COMPARE
Liquid 16000 Methyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, PVC
$300.00USD / Case of 4
Tedia MS1922-006 Methyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, PVC EW-15001-36 Mfr # MS1922-006
COMPARE
Liquid 16000 Methyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, PVC
$450.00USD / Case of 4
Tedia Methyl Alcohol, HPLC/Spectrophotometric Grade; 4 L, PVC EW-15001-38 Mfr # MS1922-073
COMPARE
Liquid 4000 Methyl Alcohol, HPLC/Spectrophotometric Grade; 4 L, PVC
$90.00USD / Each
Showing 1- 18 of 18

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat