TEDIA® n-Butyl Alcohol

High-purity solvents for life science, biotechnology, pharmaceutical, laboratory, and industrial applications.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$40.00 - $436.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Purity Grade Volume (mL) Description
Availability Pricing
Tedia BR0292-001 n-Butyl Alcohol, ACS Reagent Grade; 4 x 4 L, Amber Glass EW-15000-28 Mfr # BR0292-001
COMPARE
ACS Reagent 1000 n-Butyl Alcohol, ACS Reagent Grade; 1 L, Amber Glass
$363.00USD / Case of 4
Tedia n-Butyl Alcohol, ACS Reagent Grade; 6 x 1 L, Amber Glass EW-15000-29 Mfr # BR0292-002
COMPARE
ACS Reagent 6000 n-Butyl Alcohol, ACS Reagent Grade; 6 x 1 L, Amber Glass
$169.00USD / Case of 6
Tedia n-Butyl Alcohol, ACS Reagent Grade; 4 L, Amber Glass EW-15000-30 Mfr # BR0292-048
COMPARE
ACS Reagent 4000 n-Butyl Alcohol, ACS Reagent Grade; 4 L, Amber Glass
$104.00USD / Each
Tedia BR0292-065 n-Butyl Alcohol, ACS Reagent Grade; 1 L, Amber Glass EW-15000-31 Mfr # BR0292-065
COMPARE
ACS Reagent 1000 n-Butyl Alcohol, ACS Reagent Grade; 1 L, Amber Glass
$40.00USD / Each
Tedia n-Butyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, Amber Glass EW-15000-34 Mfr # BS1241-001
COMPARE
HPLC 16000 n-Butyl Alcohol, HPLC/Spectrophotometric Grade; 4 x 4 L, Amber Glass
$436.00USD / Case of 4
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat